missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5410003

 View more versions of this product

Catalog No. AAH5410003



Description

Description

Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

Solubility
Soluble in chloroform, ethyl acetate, and DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate
114772-40-6
MFCD06657561
YHXCWNQNVMAENQ-UHFFFAOYSA-N
tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
9906257
97%
1g
C18H19BrO2
1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr
347.252
347.24

Chemical Identifiers

114772-40-6
347.252
YHXCWNQNVMAENQ-UHFFFAOYSA-N
9906257
CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr
C18H19BrO2
MFCD06657561
1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents