missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5410006

 View more versions of this product

Catalog No. AAH5410006



Description

Description

Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

Solubility
Soluble in chloroform, ethyl acetate, and DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate
114772-40-6
MFCD06657561
Soluble in chloroform,ethyl acetate,and DMSO.
CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr
347.252
347.24
5g
C18H19BrO2
1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
YHXCWNQNVMAENQ-UHFFFAOYSA-N
tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
9906257
97%

Chemical Identifiers

114772-40-6
347.252
YHXCWNQNVMAENQ-UHFFFAOYSA-N
9906257
CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr
C18H19BrO2
MFCD06657561
1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 442-850-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents