Please login to your online account to display your discounted pricing

tert-Butyl 4-amino-1-piperidinecarboxylate 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384860010

 View more versions of this product

Catalog No. AC384860010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder, Crystals and/or Chunks
Formula Weight 200.28
Quantity 1g
Color White to pale yellow to beige
Packaging Glass bottle
Molecular Formula C10H20N2O2
MDL Number MFCD01076201
Infrared Spectrum Authentic
Synonym 4-Amino-1-BOC-piperidine
CAS 87120-72-7
Assay Percent Range 98%
Chemical Name or Material tert-Butyl 4-amino-1-piperidinecarboxylate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butyl 4-amino-1-piperidinecarboxylate
CAS 87120-72-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD01076201
Synonym 4-Amino-1-BOC-piperidine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents