missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate 98.0+%, TCI America™
Supplier: TCI America B46691G
Specifications
| tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate | |
| 133°C | |
| C14H22N4O2 | |
| 1 g | |
| RMULRXHUNOVPEI-UHFFFAOYSA-N | |
| tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate | |
| 11737525 | |
| ≥98.0% (HPLC,T) |
| 571188-59-5 | |
| Yellow | |
| MFCD11594962 | |
| tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester | |
| CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 | |
| 278.36 | |
| 278.36 | |
| Crystalline Powder |
Chemical Identifiers
| 571188-59-5 | |
| 278.36 | |
| RMULRXHUNOVPEI-UHFFFAOYSA-N | |
| 11737525 | |
| CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 |
| C14H22N4O2 | |
| MFCD11594962 | |
| tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester | |
| tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate |
Safety and Handling
TSCA : No