missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate, ≥97%, Maybridge

Chemical Identifiers

CAS 857284-20-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C15H23N3O3
Formula Weight 293.36
MDL Number MFCD08060526
Chemical Name or Material tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC43909CB SDS Fisher Scientific Company
CC43909CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43909DA SDS Fisher Scientific Company
CC43909DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

857284-20-9
100.0
C15H23N3O3
MFCD08060526
97%
96.5
97%
293.36
tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate

Specifications

293.36
100.0
96.5
MFCD08060526
857284-20-9
97%
250mg
C15H23N3O3
Amber Glass Bottle
97%
tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.