Please login to your online account to display your discounted pricing

tert-Butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate, ≥90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30046DA

 View more versions of this product

Catalog No. CC30046DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 305.42
Quantity 1g
Assay 90%
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
MDL Number MFCD08060503
CAS Min % 89.5
Packaging Amber Glass Bottle
Assay Percent Range 90%
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C17H27N3O2
CAS 681508-91-8
Chemical Name or Material Tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
CAS 681508-91-8
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C17H27N3O2
Formula Weight 305.42
MDL Number MFCD08060503
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

SDS & Safety Documents