missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Maybridge

Chemical Identifiers

CAS 158985-37-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H24N2O3
Formula Weight 292.38
MDL Number MFCD06411543
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC30009DE Fisher Scientific Company
CC30009DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30009CB SDS Fisher Scientific Company
CC30009CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30009DA SDS Fisher Scientific Company
CC30009DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

158985-37-6
100.0
C16H24N2O3
MFCD06411543
97%
96.5
97%
292.38
tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate

Specifications

158985-37-6
100.0
97%
292.38
tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
96.5
5g
C16H24N2O3
MFCD06411543
97%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.