missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Maybridge

Chemical Identifiers

CAS 197638-83-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H22N2O3
Formula Weight 290.36
MDL Number MFCD05864663
Chemical Name or Material Tert-butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC30004DA SDS Fisher Scientific Company
CC30004DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30004DE Fisher Scientific Company
CC30004DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

197638-83-8
100.0
C16H22N2O3
MFCD05864663
97%
96.5
97%
290.36
Tert-butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Specifications

197638-83-8
100.0
1g
C16H22N2O3
MFCD05864663
97%
96.5
Amber glass bottle
97%
290.36
Tert-butyl 4-(4-formylphenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning