Please login to your online account to display your discounted pricing

tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate, 90%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC67904DE

 View more versions of this product

Catalog No. CC67904DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 319.4
Quantity 5g
Assay 90%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 90%
Molecular Formula C18H25NO4
CAS 1080028-74-5
CAS Min % 89.5
MDL Number MFCD11841070
Chemical Name or Material Tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.

Specifications

Chemical Name or Material Tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate
CAS 1080028-74-5
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C18H25NO4
Formula Weight 319.4
MDL Number MFCD11841070
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents