Please login to your online account to display your discounted pricing

tert-Butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC50613DA

 View more versions of this product

Catalog No. CC50613DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 292.38
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate
Assay Percent Range 97%
CAS 910036-87-2
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD09787492
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Molecular Formula C15H24N4O2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate
CAS 910036-87-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H24N4O2
Formula Weight 292.38
MDL Number MFCD09787492
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents