Please login to your online account to display your discounted pricing

tert-Butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate
CAS 910036-87-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H24N4O2
Formula Weight 292.38
MDL Number MFCD09787492
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC50613CB

 
fisher scientific company
CC50613CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC50613DA

 
fisher scientific company
CC50613DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 292.38
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H24N4O2
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD09787492
CAS 910036-87-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.