Please login to your online account to display your discounted pricing

tert-Butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥90%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC30013DA

 View more versions of this product

Catalog No. CC30013DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 90%
Formula Weight 291.39
Quantity 1g
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS 852180-47-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 89.5
MDL Number MFCD03791213
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Molecular Formula C16H25N3O2
Assay Percent Range 90%

Specifications

Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
CAS 852180-47-3
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C16H25N3O2
Formula Weight 291.39
MDL Number MFCD03791213
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents