Please login to your online account to display your discounted pricing

tert-Butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥90%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC30013DE

 View more versions of this product

Catalog No. CC30013DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 291.39
Assay 90%
Quantity 5g
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 90%
CAS 852180-47-3
CAS Min % 89.5
Molecular Formula C16H25N3O2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03791213
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
CAS 852180-47-3
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C16H25N3O2
Formula Weight 291.39
MDL Number MFCD03791213
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.