Please login to your online account to display your discounted pricing

tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CC30709EA

 View more versions of this product

Catalog No. CC30709EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 292.38
Quantity 10g
Chemical Name or Material tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
CAS 179250-28-3
Assay Percent Range 97%
Molecular Formula C16H24N2O3

Specifications

Chemical Name or Material tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Assay Percent Range 97%
Molecular Formula C16H24N2O3
Formula Weight 292.38