missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
179250-28-3 |
Molecular Formula |
C16H24N2O3 |
Molecular Weight (g/mol) |
292.379 |
InChI Key |
MQZUQGHXDLMAKT-UHFFFAOYSA-N |
Synonym |
2-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-2-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-2-hydroxymethyl phenyl piperazine-1-carboxylate, pubchem11335, 1-boc-4-hydroxymethyl-phenyl-piperazine, 1-boc-4-2-hydroxymethyl-phenyl-piperazine, 1-2-hydroxymethyl-phenyl-piperazine-4-carboxylic, 1-t-butoxycarbonyl-4-2-hydroxymethyl-phenyl-piperazine, 4-2-hydroxymethylphenyl piperazine-1-carboxylic acid t-butyl ester |
PubChem CID |
7127802 |
IUPAC Name |
tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate |
SMILES |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CO |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
179250-28-3 |
|
292.379 |
|
2-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-2-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-2-hydroxymethyl phenyl piperazine-1-carboxylate, pubchem11335, 1-boc-4-hydroxymethyl-phenyl-piperazine, 1-boc-4-2-hydroxymethyl-phenyl-piperazine, 1-2-hydroxymethyl-phenyl-piperazine-4-carboxylic, 1-t-butoxycarbonyl-4-2-hydroxymethyl-phenyl-piperazine, 4-2-hydroxymethylphenyl piperazine-1-carboxylic acid t-butyl ester |
|
tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate |
|
C16H24N2O3 |
|
MQZUQGHXDLMAKT-UHFFFAOYSA-N |
|
7127802 |
|
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CO |
Specifications
|
179250-28-3 |
|
C16H24N2O3 |
|
2-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-2-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-2-hydroxymethyl phenyl piperazine-1-carboxylate, pubchem11335, 1-boc-4-hydroxymethyl-phenyl-piperazine, 1-boc-4-2-hydroxymethyl-phenyl-piperazine, 1-2-hydroxymethyl-phenyl-piperazine-4-carboxylic, 1-t-butoxycarbonyl-4-2-hydroxymethyl-phenyl-piperazine, 4-2-hydroxymethylphenyl piperazine-1-carboxylic acid t-butyl ester |
|
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CO |
|
292.379 |
|
292.38 |
|
97% |
|
1g |
|
MQZUQGHXDLMAKT-UHFFFAOYSA-N |
|
tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate |
|
7127802 |
|
tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate |
SDS
missing translation for 'documents'