Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AW00470DA

 View more versions of this product

Catalog No. AW00470DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 230.31
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
CAS 77279-24-4
Chemical Name or Material tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Assay Percent Range 97%
Molecular Formula C11H22N2O3

Specifications

Chemical Name or Material tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H22N2O3
Formula Weight 230.31