missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 97%

$19.98 - $19.98

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C11H22N2O3
Formula Weight 230.31
Chemical Name or Material tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AW00470DA SDS Fisher Scientific Company
AW00470DA
Amber Glass Bottle 1g Each for $19.98
AW00470DE SDS Fisher Scientific Company
AW00470DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
230.31
97%
C11H22N2O3
tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate

Specifications

230.31
Amber Glass Bottle
97%
tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
1g
77279-24-4
C11H22N2O3
97%