missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-formylphenyl)piperazine-1-carboxylate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC30704DE

 View more versions of this product

Catalog No. CC30704DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

290.36
Amber Glass Bottle
97%
174855-57-3
Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
5g
100.0
C16H22N2O3
96.5
MFCD05864664

Chemical Identifiers

174855-57-3
100.0
C16H22N2O3
MFCD05864664
96.5
97%
290.36
Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate