Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-formylphenyl)piperazine-1-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30704DA

 View more versions of this product

Catalog No. CC30704DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 290.36
Quantity 1g
Packaging Amber Glass Bottle
CAS Max % 100.0
MDL Number MFCD05864664
Molecular Formula C16H22N2O3
CAS 174855-57-3
CAS Min % 96.5
Assay Percent Range 97%
Chemical Name or Material Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate

Specifications

Chemical Name or Material Tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
CAS 174855-57-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H22N2O3
Formula Weight 290.36
MDL Number MFCD05864664