Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO07069CB

 View more versions of this product

Catalog No. MO07069CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 292.38
Quantity 250mg
Assay 97%
CAS Min % 97.0
Synonym tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate (IUPAC)
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Molecular Formula C16H24N2O3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 690632-14-5
Packaging Amber Glass Bottle
MDL Number MFCD05865139
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

Specifications

Chemical Name or Material tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
CAS 690632-14-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H24N2O3
Formula Weight 292.38
MDL Number MFCD05865139
Synonym tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate (IUPAC)
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents