missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Maybridge

Chemical Identifiers

CAS 690632-14-5
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C16H24N2O3
Formula Weight 292.38
MDL Number MFCD05865139
Synonym tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate (IUPAC)
Chemical Name or Material tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO07069CB SDS Fisher Scientific Company
MO07069CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

690632-14-5
97%
292.38
tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate (IUPAC)
97%
97.0
C16H24N2O3
MFCD05865139
tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate

Specifications

292.38
97.0
C16H24N2O3
Amber Glass Bottle
97%
97%
250mg
tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate (IUPAC)
690632-14-5
MFCD05865139
tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning