missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Maybridge

Chemical Identifiers

CAS 174855-53-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H25N3O2
Formula Weight 291.39
MDL Number MFCD04115066
Chemical Name or Material Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC30713DA SDS Fisher Scientific Company
CC30713DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30713CB SDS Fisher Scientific Company
CC30713CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

174855-53-9
100.0
C16H25N3O2
MFCD04115066
97%
96.5
97%
291.39
Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate

Specifications

291.39
100.0
174855-53-9
96.5
C16H25N3O2
97%
1g
97%
MFCD04115066
Amber Glass Bottle
Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger