Please login to your online account to display your discounted pricing

tert-Butyl (3S)-3-aminobutanoate, 95%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC337942500

 View more versions of this product

Catalog No. AC337942500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 159.23
Quantity 250mg
Color Clear colorless to light yellow
Specific Rotation Condition + 24.00 (20.00°C c=1,CHCl3)
Packaging Glass bottle
Specific Rotation +21° to +27° (20°C, 589nm) (c=1, CHCl3)
Chemical Name or Material tert-Butyl (3S)-3-aminobutanoate
Name Note 95%
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C8H17NO2
Refractive Index 1.4230 to 1.4270 (20°C, 589nm)
MDL Number MFCD00798294
CAS 161105-54-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butyl (3S)-3-aminobutanoate
Name Note 95%
CAS 161105-54-0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H17NO2
Formula Weight 159.23
MDL Number MFCD00798294
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents