missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl (3S)-3-amino-5-methylhexanoate, 95%, ACROS Organics™
Chemical Identifiers
CAS | 166023-30-9 |
---|---|
Molecular Formula | C11H23NO2 |
Molecular Weight (g/mol) | 201.31 |
MDL Number | MFCD01076260 |
InChI Key | WBOPVTYXLFEVGP-VIFPVBQESA-N |
Synonym | 3s-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoic acid,3-amino-5-methyl-, 1,1-dimethylethyl ester, 3s, hexanoic acid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3s, s-tert-butyl 3-amino-5-methylhexanoate, tert-butyl 3s-3-amino-5-methyl-hexanoate, tert-butyl 3s-3-amino-5-methylhexanoate |
PubChem CID | 7021556 |
IUPAC Name | tert-butyl (3S)-3-amino-5-methylhexanoate |
SMILES | CC(C)CC(CC(=O)OC(C)(C)C)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC337931000
|
Thermo Fisher Scientific
337931000 |
100mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
166023-30-9 | |
201.31 | |
WBOPVTYXLFEVGP-VIFPVBQESA-N | |
7021556 | |
CC(C)CC(CC(=O)OC(C)(C)C)N |
C11H23NO2 | |
MFCD01076260 | |
3s-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoic acid,3-amino-5-methyl-, 1,1-dimethylethyl ester, 3s, hexanoic acid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3s, s-tert-butyl 3-amino-5-methylhexanoate, tert-butyl 3s-3-amino-5-methyl-hexanoate, tert-butyl 3s-3-amino-5-methylhexanoate | |
tert-butyl (3S)-3-amino-5-methylhexanoate |
Specifications
Authentic | |
1.4300 to 1.4350 (20°C, 589nm) | |
Colorless to Orange-Yellow | |
166023-30-9 | |
C11H23NO2 | |
3s-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoic acid,3-amino-5-methyl-, 1,1-dimethylethyl ester, 3s, hexanoic acid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3s, s-tert-butyl 3-amino-5-methylhexanoate, tert-butyl 3s-3-amino-5-methyl-hexanoate, tert-butyl 3s-3-amino-5-methylhexanoate | |
CC(C)CC(CC(=O)OC(C)(C)C)N | |
201.31 | |
201.31 | |
95% |
Glass Bottle | |
12 | |
100mg | |
94% min. (GC) | |
MFCD01076260 | |
WBOPVTYXLFEVGP-VIFPVBQESA-N | |
tert-butyl (3S)-3-amino-5-methylhexanoate | |
7021556 | |
Liquid | |
tert-Butyl (3S)-3-amino-5-methylhexanoate, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger