Please login to your online account to display your discounted pricing

tert-Butyl (3R)-3-aminobutanoate, 95%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC337950025

 View more versions of this product

Catalog No. AC337950025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 159.23
Quantity 2.5g
Color Clear slightly yellow
Specific Rotation Condition −24° (20°C c=1,CHCl3)
Specific Rotation -21° to -27° (20°C, 589nm) (C=1, CHCl3)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Molecular Formula C8H17NO2
MDL Number MFCD00798293
Chemical Name or Material tert-Butyl (3R)-3-aminobutanoate
CAS 158849-23-1
Assay Percent Range 95%
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butyl (3R)-3-aminobutanoate
CAS 158849-23-1
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H17NO2
Formula Weight 159.23
MDL Number MFCD00798293
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents