Please login to your online account to display your discounted pricing

tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, ACROS Organics™


Quantity 100mg
Packaging Glass Bottle in Plastic Container

Manufacturer: fisher scientific acros  337921000

 View more versions of this product

Catalog No. AC337921000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 201.31
Quantity 100mg
Color Clear colorless to yellow
Packaging Glass Bottle in Plastic Container
Chemical Name or Material tert-Butyl (3R)-3-amino-5-methylhexanoate
Name Note 97%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C11H23NO2
Refractive Index 1.4300 to 1.4330 (20°C, 589nm)
MDL Number MFCD01076257
CAS 166023-29-6
Specific Rotation -11° to -15° (20°C, 589nm) (c=1, CHCl3)
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material tert-Butyl (3R)-3-amino-5-methylhexanoate
Name Note 97%
CAS 166023-29-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H23NO2
Formula Weight 201.31
MDL Number MFCD01076257
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents