Please login to your online account to display your discounted pricing

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC337880010

 View more versions of this product

Catalog No. AC337880010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Slightly Viscous Liquid
Assay ≥96.0%
Formula Weight 221.3
Color Clear colorless to yellow
Quantity 1g
Specific Rotation Condition + 17.00 (20.00°C c=1,CHCl3)
Chemical Name or Material tert-Butyl (3R)-3-amino-3-phenylpropanoate
Name Note 97%
Assay Percent Range 97%
Molecular Formula C13H19NO2
MDL Number MFCD00798309
Health Hazard 1 Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
CAS 161671-34-7
Specific Rotation +17° to +22° (20°C, 589nm) (c=1, CHCl3)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material tert-Butyl (3R)-3-amino-3-phenylpropanoate
Name Note 97%
CAS 161671-34-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H19NO2
Formula Weight 221.3
MDL Number MFCD00798309
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents