missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, Thermo Scientific™

$320.48 - $320.48

Chemical Identifiers

CAS 161671-34-7
Molecular Formula C13H19NO2
Molecular Weight (g/mol) 221.3
MDL Number MFCD00798309
InChI Key TYYCBAISLMKLMT-LLVKDONJSA-N
Synonym 1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, r-tert-butyl 3-amino-3-phenylpropanoate, tert-butyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate
PubChem CID 2733824
IUPAC Name tert-butyl (3R)-3-amino-3-phenylpropanoate
SMILES CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC337882500
SDS View Documents Promotion Details
Thermo Fisher Scientific
337882500
250mg
Each for $320.48
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

161671-34-7
221.3
TYYCBAISLMKLMT-LLVKDONJSA-N
2733824
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N
C13H19NO2
MFCD00798309
1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, r-tert-butyl 3-amino-3-phenylpropanoate, tert-butyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate
tert-butyl (3R)-3-amino-3-phenylpropanoate

Specifications

161671-34-7
97%
Authentic
Glass Bottle
250mg
TYYCBAISLMKLMT-LLVKDONJSA-N
CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N
tert-butyl (3R)-3-amino-3-phenylpropanoate
2733824
Slightly Viscous Liquid
tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%
Colorless to Yellow
C13H19NO2
MFCD00798309
1.4960 to 1.498
1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, r-tert-butyl 3-amino-3-phenylpropanoate, tert-butyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate
+ 17.00 (20.00°C c=1,CHCl3)
+ 17
221.3
221.3
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications

RUO – Research Use Only