Please login to your online account to display your discounted pricing

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%, ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  AC337882500

 View more versions of this product

Catalog No. AC337882500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color Clear colorless to yellow
Physical Form Slightly Viscous Liquid
Formula Weight 221.3
Quantity 250mg
Packaging Glass Bottle
Specific Rotation Condition + 17.00 (20.00°C c=1,CHCl3)
Specific Rotation +17° to +22° (20°C, 589nm) (c=1, CHCl3)
Infrared Spectrum Authentic
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00798309
Assay Percent Range 97%
Chemical Name or Material tert-Butyl (3R)-3-amino-3-phenylpropanoate
Molecular Formula C13H19NO2
CAS 161671-34-7
Health Hazard 1 Danger

Specifications

Chemical Name or Material tert-Butyl (3R)-3-amino-3-phenylpropanoate
Name Note 97%
CAS 161671-34-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H19NO2
Formula Weight 221.3
MDL Number MFCD00798309
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents