Alfa Aesar™ tert-Butyl 3-oxocyclobutanecarboxylate, 97%
Manufacturer:  Alfa Aesar™ H62064MD
 View more versions of this product
Catalog No. AAH62064MD
Specifications
Specifications
tert-Butyl 3-oxocyclobutanecarboxylate
145549-76-4
MFCD12827541
JINYZTGTQXDUQR-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutane-1-carboxylate
10261520
97%
250mg
C9H14O3
1-boc-cyclobutan-3-one, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, 3-tert-butyloxycarbonylcyclobutanone, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, t-butyl 3-oxocyclobutanecarboxylate, t-butyl-3-oxocyclobutanecarboxylate, tert-butyl 3-oxo-cyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, tert-butyl 3-oxocyclobutanecarboxylate, tert-butyl3-oxocyclobutanecarboxylate
CC(C)(C)OC(=O)C1CC(=O)C1
170.208
170.2
Chemical Identifiers
145549-76-4
170.208
JINYZTGTQXDUQR-UHFFFAOYSA-N
10261520
CC(C)(C)OC(=O)C1CC(=O)C1
C9H14O3
MFCD12827541
1-boc-cyclobutan-3-one, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, 3-tert-butyloxycarbonylcyclobutanone, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, t-butyl 3-oxocyclobutanecarboxylate, t-butyl-3-oxocyclobutanecarboxylate, tert-butyl 3-oxo-cyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, tert-butyl 3-oxocyclobutanecarboxylate, tert-butyl3-oxocyclobutanecarboxylate
tert-butyl 3-oxocyclobutane-1-carboxylate
Safety and Handling
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
Documents