missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, ACROS Organics™

Chemical Identifiers

CAS 898271-20-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
InChI Key XQNQFOKZUQWWDO-UHFFFAOYSA-N
Synonym 1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-boc-3-2-aminoethyl azetidine, 1-boc-3-aminoethyl azetidine, 1-boc-3-aminoethylazetidine, 1-n-boc-3-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
PubChem CID 44630603
IUPAC Name tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451232500
SDS View Documents
Thermo Fisher Scientific
AC451232500
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451230010
SDS View Documents
Thermo Fisher Scientific
AC451230010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

898271-20-0
200.28
1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-boc-3-2-aminoethyl azetidine, 1-boc-3-aminoethyl azetidine, 1-boc-3-aminoethylazetidine, 1-n-boc-3-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
C10H20N2O2
XQNQFOKZUQWWDO-UHFFFAOYSA-N
44630603
CC(C)(C)OC(=O)N1CC(C1)CCN

Specifications

250mg
94% min. (GC)
1-azetidinecarboxylic acid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-azetidinecarboxylicacid, 3-2-aminoethyl-, 1,1-dimethylethyl ester, 1-boc-3-2-aminoethyl azetidine, 1-boc-3-aminoethyl azetidine, 1-boc-3-aminoethylazetidine, 1-n-boc-3-aminoethyl azetidine, 3-2-aminoethyl-1-boc-azetidine, tert-butyl 3-2-aminoethyl azetidine-1-carboxylate, tert-butyl 3-2-aminoethyl-1-azetidinecarboxylate
CC(C)(C)OC(=O)N1CC(C1)CCN
200.28
200.28
tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate
898271-20-0
C10H20N2O2
XQNQFOKZUQWWDO-UHFFFAOYSA-N
tert-butyl 3-(2-aminoethyl)azetidine-1-carboxylate
44630603
95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS Signal Word: Danger