Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl [2-(methylthio)ethyl]carbamate, 97%

Chemical Identifiers

Chemical Name or Material tert-Butyl [2-(methylthio)ethyl]carbamate
CAS 174360-08-8
Linear Formula CH3SCH2CH2NHCO2C(CH3)3
Formula Weight 191.29
MDL Number MFCD21105444
Beilstein 7421844
Synonym [2-(Methylthio)ethyl]carbamic acid tert-butyl ester; N-Boc-2-(methylthio)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6412803

  SDS
alfa aesar™
H6412803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6412806

  SDS
alfa aesar™
H6412806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64128MD

  SDS
alfa aesar™
H64128MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.29
Chemical Name or Material tert-Butyl [2-(methylthio)ethyl]carbamate
Physical Form Liquid
Assay 97%
Linear Formula CH3SCH2CH2NHCO2C(CH3)3
MDL Number MFCD21105444
CAS 174360-08-8
Beilstein 7421844
Synonym [2-(Methylthio)ethyl]carbamic acid tert-butyl ester; N-Boc-2-(methylthio)ethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless to yellow
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.