Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl 2,2,2-trichloroacetimidate, 97%

$268.20 - $268.20

Chemical Identifiers

Chemical Name or Material tert-Butyl 2,2,2-trichloroacetimidate
CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Formula Weight 218.51
MDL Number MFCD00077410
UN Number UN1993
Beilstein 1770049
Synonym 2,2,2-Trichloroacetimidic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2203914

  SDS
alfa aesar
B2203914
25g Each for $268.20

AAB2203903

  SDS
alfa aesar™
B2203903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2203906

  SDS
alfa aesar™
B2203906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.51
Quantity 25g
Assay 97%
MDL Number MFCD00077410
UN Number UN1993
Beilstein 1770049
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density 1.222
Chemical Name or Material tert-Butyl 2, 2, 2-trichloroacetimidate
Assay Percent Range 0.97
Molecular Formula C6H10Cl3NO
CAS 98946-18-0
Storage Note 1 Moisture Sensitive

Applications
tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

Solubility
Soluble in organic solvents, cyclo hexane.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

Moisture Sensitive