missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate, TCI America™

$111.53 - $385.02

Chemical Identifiers

CAS 186020-66-6
Molecular Formula C13H26O6
Molecular Weight (g/mol) 278.345
MDL Number MFCD03791106
InChI Key KSXVEOLRERRELV-UHFFFAOYSA-N
Synonym 12-Hydroxy-4,7,10-trioxadodecanoic Acid tert-Butyl Ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate, 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic Acid tert-Butyl Ester
PubChem CID 3625858
IUPAC Name tert-butyl 3-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]propanoate
SMILES CC(C)(C)OC(=O)CCOCCOCCOCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B55871G View Documents TCI America
B55871G
1g Each for $111.53
 
B55875G View Documents TCI America
B55875G
5g Each for $385.02
 
Specifications

Chemical Identifiers

186020-66-6
278.345
KSXVEOLRERRELV-UHFFFAOYSA-N
3625858
CC(C)(C)OC(=O)CCOCCOCCOCCO
C13H26O6
MFCD03791106
12-Hydroxy-4,7,10-trioxadodecanoic Acid tert-Butyl Ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate, 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic Acid tert-Butyl Ester
tert-butyl 3-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]propanoate

Specifications

Yellow
186020-66-6
MFCD03791106
KSXVEOLRERRELV-UHFFFAOYSA-N
tert-butyl 3-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]propanoate
3625858
Clear Liquid at 20°C
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
1g
C13H26O6
12-Hydroxy-4,7,10-trioxadodecanoic Acid tert-Butyl Ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate, 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic Acid tert-Butyl Ester
CC(C)(C)OC(=O)CCOCCOCCOCCO
278.345
278.35
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No