missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butoxydiphenylchlorosilane (stabilized with CaCO3), TCI America™

$320.84 - $1,072.52

Chemical Identifiers

CAS 17922-24-6
Molecular Formula C16H19ClOSi
Molecular Weight (g/mol) 290.862
MDL Number MFCD00077672
InChI Key MSZDOMFSWXWKTK-UHFFFAOYSA-N
Synonym tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride
PubChem CID 2735650
IUPAC Name chloro-[(2-methylpropan-2-yl)oxy]-diphenylsilane
SMILES CC(C)(C)O[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14365ML View Documents TCI America
B14365ML
5mL Each for $320.84
 
B143625ML View Documents TCI America
B143625ML
25mL Each for $1,072.52
 
Specifications

Chemical Identifiers

17922-24-6
290.862
MSZDOMFSWXWKTK-UHFFFAOYSA-N
2735650
CC(C)(C)O[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
C16H19ClOSi
MFCD00077672
tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride
chloro-[(2-methylpropan-2-yl)oxy]-diphenylsilane

Specifications

112°C
5mL
C16H19ClOSi
2986
MSZDOMFSWXWKTK-UHFFFAOYSA-N
chloro-[(2-methylpropan-2-yl)oxy]-diphenylsilane
2735650
Clear Liquid at 20°C
tert-Butoxydiphenylchlorosilane (stabilized with CaCO3)
Colorless
17922-24-6
MFCD00077672
tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride
CC(C)(C)O[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
290.862
290.86
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer