Please login to your online account to display your discounted pricing

Alfa Aesar™ (tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%


Quantity 1g

Manufacturer: alfa aesar™  L1514403

 View more versions of this product

Catalog No. AAL1514403

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 376.44
MDL Number MFCD00075545
Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane
Molecular Formula C24H25O2P
Linear Formula (CH3)3CO2CCH=P(C6H5)3
UN Number UN2811
Synonym tert-Butyl (triphenylphosphoranylidene)acetate; (Carbo-tert-butoxymethylene)triphenylphosphorane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H373-H319-H317
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 146° to 150°C
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 98%
CAS 35000-38-5
Beilstein 2759875

Specifications

Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane
CAS 35000-38-5
Molecular Formula C24H25O2P
Linear Formula (CH3)3CO2CCH=P(C6H5)3
Formula Weight 376.44
MDL Number MFCD00075545
UN Number UN2811
Beilstein 2759875
Synonym tert-Butyl (triphenylphosphoranylidene)acetate; (Carbo-tert-butoxymethylene)triphenylphosphorane

Applications
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.

Accessories

Danger

GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H373-H319-H317
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.

SDS & Safety Documents