Please login to your online account to display your discounted pricing

Alfa Aesar™ (tert-Butoxycarbonylmethyl)triphenylphosphonium chloride, 98+%

$44.31 - $323.96

Chemical Identifiers

Chemical Name or Material (tert-Butoxycarbonylmethyl)triphenylphosphonium chloride
CAS 35000-37-4
Molecular Formula (CH3)3CO2CCH2P(C6H5)3Cl
Formula Weight 412.9
MDL Number MFCD00043160
Beilstein 4116684
Synonym Carbo-tert-butoxymethyl triphenylphosphonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1224506

  SDS
alfa aesar™
L1224506
5g Each for $44.31

AAL1224514

  SDS
alfa aesar™
L1224514
25g Each for $108.04

AAL1224522

  SDS
alfa aesar™
L1224522
100g Each for $323.96
Description & Specifications

Specifications

Formula Weight 412.9
Chemical Name or Material (tert-Butoxycarbonylmethyl)triphenylphosphonium chloride
Assay ≥98%
Molecular Formula (CH3)3CO2CCH2P(C6H5)3Cl
MDL Number MFCD00043160
CAS 35000-37-4
Synonym Carbo-tert-butoxymethyl triphenylphosphonium chloride
Health Hazard 1 Warning
Melting Point ≈190°C
Quantity 5g
Recommended Storage 2° to 8°C
Beilstein 4116684
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338-P337+P313-P405-P403+P233-P501a
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338-P337+P313-P405-P403+P233-P501a
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C