Please login to your online account to display your discounted pricing

Alfa Aesar™ (tert-Butoxycarbonylmethyl)triphenylphosphonium bromide, 98%

$50.34 - $50.34

Chemical Identifiers

Chemical Name or Material (tert-Butoxycarbonylmethyl)triphenylphosphonium bromide
CAS 59159-39-6
Molecular Formula C24H26BrO2P
Linear Formula (CH3)3CO2CCH2P(C6H5)3Br
Formula Weight 457.35
MDL Number MFCD00075527
Beilstein 4631405
Synonym Carbo-tert-butoxymethyl triphenylphosphonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1516506

  SDS
alfa aesar™
L1516506
5g Each for $50.34
Description & Specifications

Specifications

Formula Weight 457.35
Chemical Name or Material (tert-Butoxycarbonylmethyl)triphenylphosphonium bromide
Assay 98%
Molecular Formula C24H26BrO2P
MDL Number MFCD00075527
CAS 59159-39-6
Synonym Carbo-tert-butoxymethyl triphenylphosphonium bromide
Health Hazard 1 Warning
Linear Formula (CH3)3CO2CCH2P(C6H5)3Br
Beilstein 4631405
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point ≈178°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.