Learn More
tert-Butoxybis(dimethylamino)methane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 378120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
tert-Butoxybis(dimethylamino)methane | |
Yellow | |
50°C to 55°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4220 to 1.425 | |
MFCD00042858 | |
0.844 | |
tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane | |
HXRAMSFGUAOAJR-UHFFFAOYSA-P | |
N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine | |
79885 | |
90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane |
5815-08-7 | |
0.8440g/mL | |
41°C | |
85+% | |
C9H22N2O | |
(CH3)3COCH[N(CH3)2]2 | |
25 g | |
15, 1373 | |
Solubility in water: decomposes | |
CC(C)(C)OC(N(C)C)N(C)C | |
174.29 | |
174.29 | |
Liquid |
Chemical Identifiers
5815-08-7 | |
174.29 | |
HXRAMSFGUAOAJR-UHFFFAOYSA-P | |
79885 | |
CC(C)(C)OC(N(C)C)N(C)C |
C9H22N2O | |
MFCD00042858 | |
tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane | |
N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 227-383-9
RUO – Research Use Only