missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butoxybis(dimethylamino)methane, ACROS Organics™

$212.10 - $669.77

Chemical Identifiers

Assay Percent Range 85+%
Molecular Formula C9H22N2O
Linear Formula (CH3)3COCH[N(CH3)2]2
Formula Weight 174.29
MDL Number MFCD00042858
Chemical Name or Material tert-Butoxybis(dimethylamino)methane
Assay 90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC378120250 SDS Thermo Fisher Scientific
AC378120250
Glass bottle 25g Each for $669.77
AC378120050 SDS Thermo Fisher Scientific
AC378120050
Glass bottle 5g Each for $212.10
Specifications

Description

Alkylation reagent
Description & Specifications

Chemical Identifiers

85+%
(CH3)3COCH[N(CH3)2]2
MFCD00042858
90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane
C9H22N2O
174.29
tert-Butoxybis(dimethylamino)methane

Specifications

5815-08-7
Authentic
85+%
C9H22N2O
174.29
15, 1373
25g
90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane
0.8440g/mL
Liquid
Glass bottle
(CH3)3COCH[N(CH3)2]2
MFCD00042858
Clear pale yellow
tert-Butoxybis(dimethylamino)methane

Safety and Handling

GHS P Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning