missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Amyl Alcohol 98.0+%, TCI America™

Manufacturer:  TCI America P0059500ML

 View more versions of this product

Catalog No. P0059500ML



Specifications

Specifications

tert-Amyl Alcohol
Colorless
500mL
C5H12O
1105
MSXVEPNJUHWQHW-UHFFFAOYSA-N
2-methylbutan-2-ol
6405
Clear Liquid at 20°C
102°C
-8°C
75-85-4
MFCD00004478
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
CCC(C)(C)O
88.15
88.15
≥98.0% (GC)

Chemical Identifiers

75-85-4
88.15
MSXVEPNJUHWQHW-UHFFFAOYSA-N
6405
CCC(C)(C)O
C5H12O
MFCD00004478
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
2-methylbutan-2-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : SC0175000

TSCA : Yes

Documents