Learn More
(-)-Terpinen-4-ol, 97%, sum of enantiomers, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 270430250
Specifications
(-)-Terpinen-4-ol | |
90°C | |
Glass bottle | |
0.933 | |
− 25.00 | |
Colorless to Yellow | |
97% | |
96.0 | |
96% min. (sum of enantiomers) (GC) | |
MFCD00167108 | |
--4-terpineol, --terpinen-4-ol, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, 1r, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, r, l-4-terpineneol, l-4-terpineol, p-menth-1-en-4-ol, r--, r-1-isopropyl-4-methyl-3-cyclohexen-1-ol, r-p-menth-1-en-4-ol, unii-8vi196vs5t | |
WRYLYDPHFGVWKC-JTQLQIEISA-N | |
(1R)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol | |
5325830 | |
Liquid |
0.9330g/mL | |
Authentic | |
1.4760 to 1.4820 | |
−25° (20°C c=neat) | |
212.0°C | |
25g | |
20126-76-5 | |
100.0 | |
C10H18O | |
06, IV, 250 | |
Solubility in water: very slightly soluble., soluble in alcohol | |
CC1=CCC(CC1)(C(C)C)O | |
154.25 | |
154.25 | |
97% |
Chemical Identifiers
20126-76-5 | |
154.25 | |
WRYLYDPHFGVWKC-JTQLQIEISA-N | |
5325830 | |
CC1=CCC(CC1)(C(C)C)O |
C10H18O | |
MFCD00167108 | |
--4-terpineol, --terpinen-4-ol, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, 1r, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, r, l-4-terpineneol, l-4-terpineol, p-menth-1-en-4-ol, r--, r-1-isopropyl-4-methyl-3-cyclohexen-1-ol, r-p-menth-1-en-4-ol, unii-8vi196vs5t | |
(1R)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA