Please login to your online account to display your discounted pricing

Terpinen-4-ol, 97%, ACROS Organics™

$213.29 - $213.29

Chemical Identifiers

Chemical Name or Material Terpinen-4-ol
Name Note 97%
CAS 562-74-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00001562
Synonym 4-Carvomenthenol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC360020250

  SDS
fisher scientific acros
360020250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC360021000

  SDS
thermo fisher scientific
360021000
Glass bottle 100g Each for $213.29
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 154.25
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 06, 55
Merck Index 10, 8996
TSCA TSCA
EINECS Number 209-235-5
RTECS Number OT0175110
Density 0.929g/mL
Refractive Index 1.4760 to 1.4820 (20°C, 589nm)
Specific Rotation -1° to +1° (20°C, 589nm) (neat)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 4-Carvomenthenol
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00001562
CAS 562-74-3
Name Note 97%
Assay Percent Range 97%
Chemical Name or Material Terpinen-4-ol
Molecular Formula C10H18O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.