Please login to your online account to display your discounted pricing

Terfenadine, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  348820050

 View more versions of this product

Catalog No. AC348820050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98%
Physical Form Powder
Formula Weight 471.68
Quantity 5g
Color White
EINECS Number 256-710-8
Melting Point 145° to 152°C
RTECS Number TM4969000
Merck Index 15, 9306
Chemical Name or Material Terfenadine
Infrared Spectrum Authentic
Molecular Formula C32H41NO2
MDL Number MFCD00079622
CAS 50679-08-8
Packaging Glass bottle

Specifications

Chemical Name or Material Terfenadine
Assay ≥98%
Molecular Formula C32H41NO2
Formula Weight 471.68
MDL Number MFCD00079622