Please login to your online account to display your discounted pricing

Terfenadine, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Terfenadine
Assay 98% min.
Molecular Formula C32H41NO2
Formula Weight 471.68g/mol
MDL Number MFCD00079622
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC348820050

  SDS
fisher scientific acros
348820050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 145° to 152°C
Quantity 5g
Chemical Name or Material Terfenadine
CAS 50679-08-8
Physical Form White powder
Assay 98% min.
Molecular Formula C32H41NO2
Formula Weight 471.68g/mol
MDL Number MFCD00079622
Merck Index 15, 9306
RTECS Number TM4969000
EINECS Number 256-710-8