Please login to your online account to display your discounted pricing

Terephthaloyl Chloride, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  153100050

 View more versions of this product

Catalog No. AC153100050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Flakes, Crystals, and or Chunks
Formula Weight 203.02
Quantity 5g
Color White
EINECS Number 202-829-5
TSCA TSCA
Melting Point 79° to 83°C
RTECS Number WZ1797000
Beilstein 09, 844
Chemical Name or Material Terephthaloyl chloride
Name Note 99+%
Infrared Spectrum Authentic
Linear Formula C6H4(COCl)2
Synonym 1, 4-Benzenedicarbonyl chloride
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99+%
Molecular Formula C8H4Cl2O2
MDL Number MFCD00000693
CAS 100-20-9

Specifications

Chemical Name or Material Terephthaloyl chloride
Name Note 99+%
CAS 100-20-9
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H4Cl2O2
Linear Formula C6H4(COCl)2
Formula Weight 203.02
MDL Number MFCD00000693
Synonym 1, 4-Benzenedicarbonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.

SDS & Safety Documents