Please login to your online account to display your discounted pricing

Alfa Aesar™ Terephthalic acid mono(2-bromoethyl) ester, 98+%

$358.93 - $358.93

Chemical Identifiers

Chemical Name or Material Terephthalic acid mono(2-bromoethyl) ester
CAS 173550-97-5
Molecular Formula C10H9BrO4
Formula Weight 273.08g/mol
MDL Number MFCD01075655
Synonym 2-Bromoethyl hydrogen terephthalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1370806

 
alfa aesar™
L13708-06
5g Each for $358.93
Description & Specifications

Specifications

Quantity 5g
Melting Point 173°C
Quantity 5g
Chemical Name or Material Terephthalic acid mono(2-bromoethyl) ester
CAS 173550-97-5
Assay ≥98%
Molecular Formula C10H9BrO4
Formula Weight 273.08g/mol
MDL Number MFCD01075655
Synonym 2-Bromoethyl hydrogen terephthalate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.