Learn More
Terephthaldehyde monodiethylacetal, 97%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC294630250
Specifications
Terephthaldehyde monodiethylacetal | |
>112°C | |
Glass bottle | |
1.04 | |
89°C to 93°C (7.0mmHg) | |
25g | |
81172-89-6 | |
C12H16O3 | |
MFCD00010217 | |
Solubility in water: decomposes. Other solubilities: soluble in alcohol,chlorinated solvents and,toluene | |
CCOC(C1=CC=C(C=C1)C=O)OCC | |
208.26 | |
208.26 | |
97% |
1.0400g/mL | |
Authentic | |
1.5050 to 1.5075 | |
0.1 % triethylamine | |
Brown to White | |
97% | |
96% min. (GC) | |
CHOC6H4CH(OC2H5)2 | |
4-diethoxymethy benzaldehyde, 4-diethoxymethyl benzaldehyde, 4-diethoxymethyl-benzaldehyde, 4-diethyoxymethyl benzaldehyde, acmc-209tbs, benzaldehyde, 4-diethoxymethyl, pubchem22621, terephthalaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, terephthaldehyde mono diethyl acetal | |
HTMXMFARWHNJDW-UHFFFAOYSA-N | |
4-(diethoxymethyl)benzaldehyde | |
595993 | |
Powder or Crystalline Powder |
Chemical Identifiers
81172-89-6 | |
208.26 | |
HTMXMFARWHNJDW-UHFFFAOYSA-N | |
595993 | |
CCOC(C1=CC=C(C=C1)C=O)OCC |
C12H16O3 | |
MFCD00010217 | |
4-diethoxymethy benzaldehyde, 4-diethoxymethyl benzaldehyde, 4-diethoxymethyl-benzaldehyde, 4-diethyoxymethyl benzaldehyde, acmc-209tbs, benzaldehyde, 4-diethoxymethyl, pubchem22621, terephthalaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, terephthaldehyde mono diethyl acetal | |
4-(diethoxymethyl)benzaldehyde |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning