Learn More
Terephthalaldehyde mono(diethyl acetal), 97%, stab., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1541306
Description
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.
Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.
Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Specifications
Terephthalaldehyde mono(diethyl acetal) | |
81172-89-6 | |
89°C to 93°C (7 mmHg) | |
C12H16O3 | |
MFCD00010217 | |
4293872 | |
4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde | |
HTMXMFARWHNJDW-UHFFFAOYSA-N | |
4-(diethoxymethyl)benzaldehyde | |
595993 | |
98% |
stabilized | |
1.047 | |
>110°C (230°F) | |
1.507 | |
5 g | |
Air Sensitive | |
soluble in alcohol,chlorinated solvents and toluene. Decomposes in water. | |
CCOC(OCC)C1=CC=C(C=O)C=C1 | |
208.26 | |
208.26 |
Chemical Identifiers
81172-89-6 | |
208.26 | |
HTMXMFARWHNJDW-UHFFFAOYSA-N | |
595993 | |
CCOC(OCC)C1=CC=C(C=O)C=C1 |
C12H16O3 | |
MFCD00010217 | |
4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde | |
4-(diethoxymethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only