missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terephthalaldehyde mono(diethyl acetal), 98%, stab.

Manufacturer:  Alfa Aesar™ A1541336

 View more versions of this product

Catalog No. AAA1541336



Description

Description

Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Specifications

Specifications

Terephthalaldehyde mono(diethyl acetal)
89°C to 93°C (7mmHg)
stabilized
C12H16O3
4293872
HTMXMFARWHNJDW-UHFFFAOYSA-N
4-(diethoxymethyl)benzaldehyde
595993
98%
>110°C (230°F)
500g
81172-89-6
MFCD00010217
4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
CCOC(C1=CC=C(C=C1)C=O)OCC
208.257
208.26

Chemical Identifiers

81172-89-6
208.257
HTMXMFARWHNJDW-UHFFFAOYSA-N
595993
CCOC(C1=CC=C(C=C1)C=O)OCC
C12H16O3
MFCD00010217
4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
4-(diethoxymethyl)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents